Learn More
Thermo Scientific™ Oxoid™ Baird Parker Medium Cm275 Oxoid
Brand: Thermo Scientific™ Oxoid™ CM0275T
1841.16 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Straw powder | |
Egg Yolk Tellurite Emulsion | |
6.6 to 7.0 | |
5 kg |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Baird Parker Medium Cm275 Oxoid >
Spot an opportunity for improvement?Share a Content Correction