missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bandmate™ Automated Western Blot Processor

The Bandmate Automated Western Blot Processor is a benchtop instrument that automatically performs the hands-on steps of western blot processing, including blocking, washing, and antibody incubations.

Brand:  Invitrogen™ BW1000

7204.80 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.


8357.23 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 17102886

  • £9860.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications
Show More
Product Suggestions

Product Suggestions

Videos

Videos

  • Video Player is loading.
    Current Time 0:00
    Duration 6:57
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 6:57
     
    1x

    How to Use the Invitrogen Bandmate Automated Western Blot Processor EN

SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Bandmate™ Automated Western Blot Processor > Automated Western Blot Processor

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.