missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride Dihydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Description
Barium Chloride Dihydrate, Extra Pure, SLR

Specifications
Specifications
Boiling Point | 1560°C |
Melting Point | 962°C |
Color | White |
pH | 5 |
Physical Form | Solid |
Quantity | 5 kg |
Formula Weight | 244.28g/mol |
Packaging | Plastic powder jar |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Safety and Handling

Product Identifier
- Barium chloride dihydraat
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Barium Chloride Dihydrate, Extra Pure, SLR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction