missing translation for 'onlineSavingsMsg'
Learn More
Barium Chloride R1 EU Pharmacopoeia, Fisher Chemical™ Product Code.: 12960364
compliance-icons

Barium Chloride R1 EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12960364
1000 mL, Bottle
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
This item is not returnable. View return policy

Product Code. 12960364

Brand: Fisher Chemical RP 028 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Quantity 1000 mL
Packaging Bottle
Chemical Name or Material Barium chloride solution R1
Grade EU Ph Ref: 1009301
compliance-icons
Product Identifier
  • Barium chloride Solution R1.
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash hands, forearms, andface thoroughly after handling.
Supplemental information
  • MIXTURE LIST-Contains : barium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium Chloride R1 EU Pharmacopoeia, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.