missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride R2 EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
CAS | 101-27-9 |
Quantity | 1000 mL |
Molecular Formula | C11H9Cl2NO2 |
Synonym | barban, barbamate, barbane, carbine, carbyne, chlorinat, carbin, karbin, neoban, carbyne herbicide |
InChI Key | MCOQHIWZJUDQIC-UHFFFAOYSA-N |
SMILES | C1=CC(=CC(=C1)Cl)NC(=O)OCC#CCCl |
IUPAC Name | 4-chlorobut-2-ynyl N-(3-chlorophenyl)carbamate |
Molecular Weight (g/mol) | 258.098 |
PubChem CID | 7551 |
ChEBI | CHEBI:33421 |
Show More |
Safety and Handling
Product Identifier
- Barium chloride solution R2. European Pharmacopoeia Ref: 1009302. (RP 029)
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- EUH210-Safety data sheet available on request.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'