missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1560°C |
Melting Point | 963°C |
Color | White |
Physical Form | Beads |
Quantity | 25 g |
Assay Percent Range | 99.99% (Metals basis) |
Formula Weight | 208.25 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 3.9 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y18F004 | Certificate of Analysis | 04/10/2023 | 013686.06,013686.14 |
U31J054 | Certificate of Analysis | 22/09/2023 | 013686.06,013686.14 |
G07X008 | Certificate of Analysis | 07/12/2021 | 013686.06,013686.14 |
B29Y035 | Certificate of Analysis | 07/12/2021 | 013686.06,013686.14 |
L23G08 | Certificate of Analysis | 07/12/2021 | 013686.06,013686.14 |
Safety and Handling

Product Identifier
- Barium chloride, ultra dry
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contains : Barium chloride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Barium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction