missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium chromate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Barium chromate has been found to be useful in many capacities. often the compound is used as a carrier for the chromium ions. is an oxidizing agent, making it useful as a burn rate modifier in pyrotechnic compositions. It is especially useful in delay compositions such as delay fuses. It is also used in the composition of a catalyst for alkane dehydration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Yellow |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN1479 |
Merck Index | 14,972 |
Solubility Information | Insoluble in water. Soluble in strong acids. |
Formula Weight | 253.33 |
Percent Purity | 99.998% |
Grade | Puratronic™ |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'