Learn More
Barium fluoride, 99%, Thermo Scientific Chemicals
Description
It finds applications in ceramic flux, glass manufacturing, metallurgy, electronics and dry-film lubricants. It is transparent from ultraviolet to infrared. Barium fluoride is usually employed as a scintillator material, and useful as a transmission window material for IR spectroscopy, mostly used for analysis of fuel oil samples. Of all the optical fluorides, it is the most resistant to high-energy radiation. It is used as a preopacifying agent, and in the production of enamel, welding agents, and glazing frits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 2260°C |
Melting Point | 1353°C |
Color | Colorless |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99% |
Solubility Information | Soluble in ethanol,methanol,and aqueous solutions of ammonium chloride. Slightly soluble in water. |
IUPAC Name | barium(2+) difluoride |
Formula Weight | 175.36 |
Odor | Odorless |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U01K008 | Certificate of Analysis | 20/08/2024 | 012338.30,012338.A1,012338.14 |
M19K029 | Certificate of Analysis | 24/01/2024 | 012338.30,012338.A1,012338.14 |
S09J005 | Certificate of Analysis | 24/10/2023 | 012338.30,012338.A1,012338.14 |
S27J027 | Certificate of Analysis | 24/10/2023 | 012338.30,012338.A1,012338.14 |
U24J027 | Certificate of Analysis | 21/09/2023 | 012338.30,012338.A1,012338.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.