missing translation for 'onlineSavingsMsg'
Learn More
Barium iodide hydrate, 98%, for analysis, Thermo Scientific Chemicals Product Code.: 10431061

Barium iodide hydrate, 98%, for analysis, Thermo Scientific Chemicals

Product Code. 10431061
50 g, Glass bottle
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10431061

Brand: Thermo Scientific Acros 208000500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 740.0°C
CAS 85017-89-6
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 97.5% min. (on dry substance) (Argentometry)
Molecular Formula BaI2
Linear Formula BaI2·xH2O
MDL Number MFCD00149158
Merck Index 14, 980
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol and acetone
InChI Key SGUXGJPBTNFBAD-UHFFFAOYSA-L
SMILES [I-].[I-].[Ba++]
IUPAC Name barium(2+) diiodide
Molecular Weight (g/mol) 391.14
Formula Weight 391.14
Percent Purity 98%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Barium iodide hydrate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467716 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number208000500
Lot NumberA0431956 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number208000500
Lot NumberA0386172 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number208000500
Lot NumberA0417839 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number208000500
compliance-icons
Product Identifier
  • Barium iodide hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium iodide hydrate, 98%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.