missing translation for 'onlineSavingsMsg'
Learn More
Barium manganate, tech. 90%, Thermo Scientific Chemicals Product Code.: 11307956

Barium manganate, tech. 90%, Thermo Scientific Chemicals

Product Code. 11307956
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11307956

Brand: Thermo Scientific Alfa Aesar L13172.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-35-1
Molecular Formula BaMnO4
Molecular Weight (g/mol) 256.26
MDL Number MFCD00011514
InChI Key ZZCNKSMCIZCVDR-UHFFFAOYSA-N
Synonym barium manganate, ba.mno4, acmc-1bdpw, manganic acid h2mno4 , barium salt 1:1, barium 2+ ion manganate, barium 2+ ; dioxido dioxo manganeseShow MoreShow Less
PubChem CID 3084030
IUPAC Name barium(2+);dioxido(dioxo)manganese
SMILES [Ba++].[O-][Mn]([O-])(=O)=O

Specifications

Quantity 25 g
UN Number UN3087
Merck Index 14,981
Formula Weight 256.28
Percent Purity 90%
Grade Technical
Sensitivity Moisture sensitive
Density 4.85
Chemical Name or Material Barium manganate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10232189 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL13172.06,L13172.14
Lot Number10241552 Certificate TypeCertificate of Analysis Date24/06/2022 Catalog NumberL13172.06,L13172.14
Lot Number10127221 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL13172.06,L13172.14
Lot Number10232189 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13172.06,L13172.14
Lot NumberI1301A Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13172.06,L13172.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium manganate, tech. 90%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.