missing translation for 'onlineSavingsMsg'
Learn More
Barium peroxide, 95%, Thermo Scientific Chemicals Product Code.: 10123431

Barium peroxide, 95%, Thermo Scientific Chemicals

Product Code. 10123431
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10123431

Brand: Thermo Scientific Acros 350410250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 450.0°C
CAS 1304-29-6
Color Beige
CAS Min % 94.0
CAS Max % 100.0
Physical Form Powder
Quantity 25 g
Assay Percent Range 94% min. (Iodometry)
Molecular Formula BaO2
Linear Formula BaO2
MDL Number MFCD00003454
Merck Index 15,985
Synonym barium peroxide, barium dioxide, barium binoxide, barium superoxide, barium oxide, per, bario perossido di, bariumperoxid german, bariumperoxyde dutch, dioxyde de baryum french, barium peroxide ba o2
Solubility Information Solubility in water: 0.5g/L
InChI Key ZJRXSAYFZMGQFP-UHFFFAOYSA-N
SMILES [O-][O-].[Ba+2]
IUPAC Name barium(2+);peroxide
Molecular Weight (g/mol) 169.33
PubChem CID 14773
Formula Weight 169.33
Percent Purity 95%
Packaging Plastic bottle
Chemical Name or Material Barium Peroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467357 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number350415000,350410250,350410025
Lot NumberA0455309 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number350415000,350410250,350410025
Lot NumberA0402380 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number350415000,350410250,350410025
Lot NumberA0326139 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number350415000,350410250,350410025
Lot NumberA0435001 Certificate TypeCertificate of Analysis Date05/10/2021 Catalog Number
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Barium peroxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium peroxide, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.