missing translation for 'onlineSavingsMsg'
Learn More
Barium Sulfate, U.S.P., J.T. Baker™ Product Code.: 15577884

Barium Sulfate, U.S.P., J.T. Baker™

Product Code. 15577884
500 g, Poly Bottle
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15577884

Brand: Avantor J.T.Baker 104001

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Specifications

CAS 7727-43-7
Quantity 500 g
Molecular Formula BaO4S
MDL Number MFCD00003455
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
SMILES [Ba++].[O-]S([O-])(=O)=O
IUPAC Name barium(2+);sulfate
Molecular Weight (g/mol) 233.38
PubChem CID 24414
Formula Weight 233.39
Grade USP
Packaging Poly Bottle
Specific Gravity 1L = 4.50kg
Chemical Name or Material Barium Sulfate
Show More Show Less
  • Multiple Manufacturers
    Use code 24095 to receive this promotion.
    Promo Code: 24095

    Expires2025-04-15

    Days

    13

    Hours

    21

    Minutes

    23

    Seconds

    7
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 17/03/2025 and 15/04/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.

    Terms & Conditions Learn More
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium Sulfate, U.S.P., J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.