missing translation for 'onlineSavingsMsg'
Learn More
Barium titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11399219

Barium titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11399219
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11399219

Brand: Thermo Scientific Alfa Aesar 088266.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vacuum deposition

Barium titanium oxide is used for vacuum deposition. It is also used in electronic ceramics, PTC thermistor, the configuration of the capacitors and as a raw material and intermediate in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
Synonym Barium titanate
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifications

Melting Point 1620°C
Physical Form Pieces
Quantity 25 g
Merck Index 14,1000
Solubility Information Insoluble in water.
Formula Weight 233.19
Percent Purity 99.9%
Odor Odorless
Density 5.85 g/mL
Assay (metals basis)
Chemical Name or Material Barium titanium oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW25F048 Certificate TypeCertificate of Analysis Date18/11/2022 Catalog Number088266.22,088266.14
Lot NumberM02F025 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088266.22,088266.14
Lot NumberJ29M19 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088266.22,088266.14
Lot NumberK22B25 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088266.22,088266.14
Lot NumberF08S010 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number088266.22,088266.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.