Learn More
Brannan™ Batch Sampling Incubator Thermometer with Calibration Certificate
Precision test incubator thermometers sealed into a 30 mL polypropylene bottle containing a heat transfer medium, ideally suited to monitor the temperature of samples in a controlled storage. Complete with calibration.
Brand: Brannan™ 22/105/2 + 49/503/0
Description
- Non hazardous LO-tox™ filling
- Bottle should be filled with monpropylene glycol or other heat transfer medium before use
- Temperature measurement unaffected by short term fluctuations
- Temperature range: 18°C to 50°C
- Supplied with calibration and statement of accuracy
- Made in UK
Specifications
Customer to fill with monopropylene glycol or other heat transfer medium before use | |
ISO 17025 UKAS | |
5.71 in. | |
Polypropylene bottle and glass thermometer | |
Glass | |
1.9577 oz. | |
0.14 in. | |
+50°C | |
+18°C | |
+18°C to +50°C | |
Incubator | |
± 0.5°C | |
None |
20°C, 35°C, 50°C | |
Monitor temperature of samples in a controlled storage | |
14.5 cm | |
14.5 cm | |
No | |
55.5 g | |
122°F | |
64°F | |
64°F to 122°F | |
3-point UKAS | |
None | |
1.0°C | |
3.5 cm |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.