missing translation for 'onlineSavingsMsg'
Learn More

BD Calcium Assay Kit

For homogeneous measurement of intracellular Calcium changes

Brand:  BD 640176

Additional Details : Weight : 0.48200kg

 View more versions of this product

Product Code. 15806289

  • £935.00 / 1 set
Estimated Shipment: 18-10-2024
to see stock.
Add to basket

Includes: Calcium Indicator, 10X Signal Enhancer

Description

Description

  • Assay involves only one dye addition step and does not require washing
  • Allows easy implementation in high throughput environment

Specifications

Specifications

-20°C
1 vial calcium indicator,10mL signal enhancer (10X, lyophilized), 90mL calcium assay buffer 1X
Detection Assay Kit
For homogeneous measurement of intracellular calcium changes
10 Preps
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research use only