missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Calcium Assay Kit
For homogeneous measurement of intracellular Calcium changes
£935.00 - £5560.00
Specifications
Content And Storage | -20°C |
---|---|
For Use With (Application) | For homogeneous measurement of intracellular calcium changes |
Type | Detection Assay Kit |
Includes: Calcium Indicator, 10X Signal Enhancer
Product Code | Brand | Includes | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Quantity | Price | Quantity & Availability | ||||
15806289
|
BD
640176 |
1 vial calcium indicator,10mL signal enhancer (10X, lyophilized), 90mL calcium assay buffer 1X | 10 Preps |
£935.00
1 set |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
15826289
|
BD
640178 |
10 vials calcium indicator,100mL signal enhancer (10X, lyophilized) | 100 Preps |
£5560.00
1 set |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
Description
- Assay involves only one dye addition step and does not require washing
- Allows easy implementation in high throughput environment
Specifications
-20°C | |
For homogeneous measurement of intracellular calcium changes | |
Detection Assay Kit |
Research use only