Learn More
BD Calcium Assay Kit
For homogeneous measurement of intracellular Calcium changes
Brand:  BD 640178
Additional Details : Weight : 0.47500kg
 View more versions of this product
Product Code. 15826289
  • £5560.00 / 1 set
Estimated Shipment: 18-10-2024
to see stock.
Add to basket
Includes: Calcium Indicator, 10X Signal Enhancer
Description
Description
  • Assay involves only one dye addition step and does not require washing
  • Allows easy implementation in high throughput environment
Specifications
Specifications
-20°C
10 vials calcium indicator,100mL signal enhancer (10X, lyophilized)
Detection Assay Kit
For homogeneous measurement of intracellular calcium changes
100 Preps
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research use only