Learn More
BD Difco™ Haemophilus influenzae Serotyping Antisera
Use in slide agglutination tests for serotyping Haemophilus influenzae
Brand: BD 227891
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
- Lyophilized
- Rehydrate to 1mL
- Type: Antisera
- Shelf Life: 965 days

Specifications
Refrigerate, store at 2°C to 8°C | |
Haemophilus influenzae Antiserum Type c | |
Antiserum | |
Use in slide agglutination tests for serotyping Haemophilus influenzae |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- 1ML HINFLUENZAE ANTISERUM TYPE C (+2-8C)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium azide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.