Learn More
BD Difco™ Haemophilus influenzae Serotyping Antisera
Use in slide agglutination tests for serotyping Haemophilus influenzae
£157.00 - £647.00
Specifications
Content And Storage | Refrigerate, store at 2°C to 8°C |
---|---|
Product Type | Antiserum |
For Use With (Application) | Use in slide agglutination tests for serotyping Haemophilus influenzae |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Product Code | Brand | Product Type | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Description | Price | Quantity & Availability | ||||
11722734
|
BD
222371 |
Antiserum | Haemophilus influenzae Antiserum Poly |
£647.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11788333
|
BD
222361 |
Antiserum | Haemophilus influenzae Antiserum Type b |
£551.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11732734
|
BD
222501 |
Antiserum | Haemophilus influenzae Antiserum Type a |
£463.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11742874
|
BD
227891 |
Antiserum | Haemophilus influenzae Antiserum Type c |
£157.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11752874
|
BD
227901 |
Antiserum | Haemophilus influenzae Antiserum Type d |
£157.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11762874
|
BD
227911 |
Antiserum | Haemophilus influenzae Antiserum Type e |
£157.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11782874
|
BD
227921 |
Antiserum | Haemophilus influenzae Antiserum Type f |
£157.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Lyophilized
- Rehydrate to 1mL
- Type: Antisera
- Shelf Life: 965 days
Specifications
Refrigerate, store at 2°C to 8°C | |
Antiserum | |
Use in slide agglutination tests for serotyping Haemophilus influenzae |
Safety and Handling
missing translation for 'shelfLife' : 965 Days
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium Azide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.