Learn More
BD Lineage Antibody Cocktail
Designed for flow cytometric identification of hematopoietic progenitors in mouse bone marrow
Brand: BD 558074
837.08 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: APC Mouse Lineage Antibody Cocktail, APC Mouse Lineage Isotype Control Cocktail
Description
React with cells from the major hematopoietic lineages, such as T lymphocytes, B lymphocytes, monocytes/macrophages, NK cells, erythrocytes, and granulocytes
- Contains equivalent concentrations of isotype-matched negative-control immunoglobulin
- Additional fluorochrome-labeled reagents may be combined to further characterize hematopoietic progenitor subpopulations
- Used to deplete cells bearing hematopoietic lineage markers by flow cytometric or immunomagnetic sorting
- 20μl of antibody or isotype control cocktail is sufficient to stain each sample of 106 leukocytes
- Excited by laser lines from 595 to 647nm, and its emission is collected in a detector for fluorescence wavelengths between 640 and 680nm
Flow Cytometry

Specifications
Flow Cytometry | |
APC | |
Affinity Purified | |
RUO |
Monoclonal | |
Mouse | |
1 Each | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research use only