Learn More
BD Mouse Th1/Th17 Phenotyping Kit
Simultaneously view TH17 and TH1 in mouse model systems
Brand: BD 560759
620.51 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Mouse Th1/Th17 Phenotyping Cocktail (CD4 PerCP-Cy™ 5.5, IL-17A PE, Mouse IFN-γ FITC), BD Cytofix™ Fixation Buffer, BD Perm/Wash™ Buffer, BD GolgiStop™ Protein Transport Inhibitor
Description
- Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
- Can be used to analyze ex vivo lymphoid cell samples or to monitor T helper cell differentiation
- Ensures indentification and characterization of the nature of the T helper cell types
- Allow for staining of surface CD4 along with intracellular factors
Specifications
1 Kit |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Product Title
BD Mouse Th1/Th17 Phenotyping Kit > 1 Kit (50 Tests)
For research use only. Not for diagnostic or therapeutic procedures.
Spot an opportunity for improvement?Share a Content Correction