missing translation for 'onlineSavingsMsg'
Learn More

BD T Cell Kit Lyophilized Cells

Intended to serve as biological controls for antibody staining in BD Phosflow studies designed to analyze cell signaling events associated with T-cell activation

Brand:  BD 560760

523.02 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15860469

  • £531.00 / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket

Includes: 5 vials human treated and untreated control cells
This item is not returnable. View return policy

Description

Description

Intracellular Staining, Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Flow Cytometry
10 Each
RUO
Store unopened vials of lyophilized cells at 2 - 8°C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD T Cell Kit Lyophilized Cells > T Cell Kit Lyophilized Cells

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only