missing translation for 'onlineSavingsMsg'
Learn More
Pal International™ Non-woven Fabric Beard Masks Product Code.: 12353996
Add to basket

Pal International™ Non-woven Fabric Beard Masks

Product Code. 12353996
White
Click to view available options
Color:
Blue
White

Product Code. 12353996

Brand: Pal International™ N43110HP

Edge
Add to basket
Add to basket

Soft spun-bond nonwoven fabric beard masks feature comfortable elastic ear-loops and provide effective barrier protection.

  • Latex-free
  • Soft spun-bond non-woven fabric
  • Soft elastic ear-loops
  • One size fits all

Specifications

Type General Purpose Beard Cover
Size Universal
Material Polypropylene
Color White
Disposable Single-use
Latex Free Yes
For Use With (Application) General Maintenance, Food Industry
Packaging Quantity 1000/Pk.
Length (Metric) 120 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pal International™ Non-woven Fabric Beard Masks >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.