Learn More
Novus Biologicals™ Beclin 1 Antibody Pack
For Research applications
Brand:  Novus Biologicals™ NB910-95610
Product Code. 18257623
  • £627.00 / Pack of 4
Explore more special offers
Includes:
  • 1.NB110-87318: Beclin 1/ATG6 Antibody
  • 2.NB500-249: Beclin 1/ATG6 Antibody
  • 3.NB500-266: Beclin 1/ATG6 Antibody
Specifications
Beclin 1
Unconjugated
See individual datasheets
BECN1
3 Vials
8678
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin), KnockDown
Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30
See individual datasheets
Cancer, mTOR Pathway
1.NB110-87318: Beclin 1/ATG6 Antibody
2.NB500-249: Beclin 1/ATG6 Antibody
3.NB500-266: Beclin 1/ATG6 Antibody
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Novus Biologicals™ Beclin 1 Antibody Pack > 3 vials