missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Clear View Fume Hood

Safely vent noxious or potentially hazardous fumes and dust with the Bel-Art™.

Brand:  Bel-Art™ F50020-2010

2422.93 GBP valid until 2025-04-30
Use promo code "24235" to get your promotional price.


 View more versions of this product

Product Code. 17207719

  • £2550.45 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Laboratory, classroom or industrial
This item is not returnable. View return policy

Description

Description

  • Versatile 3-position sash
  • Clear polycarbonate provides optimum visibility and durability
  • 500201010, Top sitting: 6 in. O.D. vent
  • O.D.: 24D x 25.8W x 26.12 in. H (61 x 65.7 x 66.4 cm)
  • I.D.: 20D x 24W x 24 in. H (50.8 x 61 x 61 cm)
  • Internal Space: 11.67 cu. ft. (0.18 m)
  • 500202010, Two Top sitting: 6 in. O.D. vent
  • O.D.: 24D x 43.8W x 26.1 in. H (61 x 111.4 x 66.4cm)
  • I.D.: 20D x 42W x 24 in. H (50.8 x 106.7 x 61 cm)
  • Internal Space: 11.67 cu. ft. (0.18 m)
Specifications

Specifications

Fume Hood
Unfiltered
Polycarbonate
61 cm
26 in.
43.87 in.
Uncoated
None
24 in.
111.4 x 61.0 x 66.3 cm
Laboratory, Classroom
111.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Discover exclusive monthly offers on scientific equipment to enhance your laboratory's capabilities.
    Promo Code: 24235

    Expires2025-04-30

    Days

    27

    Hours

    15

    Minutes

    14

    Seconds

    3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Clear View Fume Hood > 2 top-sitting 6 in. O.D. vent opening; I.D.: 20D x 42W x 24 in. H; O.D.: 24D x 43.8W x 26.1 in. H

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.