missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Sterileware™ V-shaped Sampling Spatula, Sterile

Ideal for every type of powdered or granulated materials. Spatulas are ideal for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.

Brand:  Bel-Art™ H36930-0000

Product Code. 11897822

  • £317.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Ideal for every type of powdered or granulated materials. Spatulas are ideal for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.

  • Time-honored V-shape makes transferring powdered, flaked, crystalline or granular media a breeze.
  • Long, 14 in. spatula (16-241-210) is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 0.75 in. (18 mm).
  • Sterileware (16-241-210, 14-385-20) available in 9 and 14 in. models.
  • High-impact polystyrene.
  • Individually packaged.
  • Sterilized by gamma irradiation.
  • Lot stamped, shipped with Certificate of Processing.
  • Nonsterile Model (03-421-149) 9 in. spatula.
Specifications

Specifications

Sterile
Pointed End
Sterileware™ Sampling Spatula
Ideal for every type of powdered or granulated material, perfect for taking sterile samples during process quality inspections and for piercing Plastic-lined Paper bags or similar package
0.75 in.
9 x 0.75 in. (22.9 x 1.9 cm)
2.99 in.
Polystyrene
Not Autoclavable
Single-use
Containers, bags and drums
9 in.
1.9 cm
6 in. L V-shaped channel
7.6 cm
22.9 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Sterileware™ V-shaped Sampling Spatula, Sterile > Sterile; Overall L: 9 in. (23cm)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.