missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Sterileware™ Sampling Spatula, 14 in.

Ideal for every type of powdered or granulated material. Spatulas are ideal for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.

Brand:  Bel-Art™ H36930-0050

Product Code. 13442489

  • £150.00 / Pack of 50
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Time-honored V-shape makes transferring powdered, flaked, crystalline or granular media a breeze
Long, 14 in. spatula is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 0.75 in. (18mm)

  • 14 in. length
  • High-impact polystyrene
  • Individually packaged
  • Sterilized by gamma irradiation
  • Lot stamped, shipped with Certificate of Processing
  • Sterile 9-in. model is available, 14-358-20
  • Nonsterile 9-in. model: 03-421-149

 

Specifications

Specifications

Sterile
Pointed End
Single-use
Tall bottles or vessels
10 in.
4 in.
Polystyrene
Not Autoclavable
23 mL
Sampling Spatula with V-shaped channel
Ideal for every type of powdered or granulated material, perfect for taking sterile samples during process quality inspections, dislodge caked or crystallized reagents from deep within tall bottles or vessels
14 in.
10.2 cm
35.6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Sterileware™ Sampling Spatula, 14 in. > Sterile; Overall L: 14 in.(35.6cm)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.