Learn More
Bel-Art™ Transpette™ Eight Channel Transfer Pipettor
For washing plates, feeding, and supernatant aspiration from microwell plates
Brand:  Bel-Art™ F37873-0001
 View more versions of this product
Product Code. 10075210
  • £327.31 / Pack of 100
Estimated Shipment: 21-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For washing plates, feeding, and supernatant aspiration from microwell plates
  • 8-Channel with 600μL capacity per channel
  • Made of translucent plastic
  • Discard after hot radioactive uses to avoid radiation contamination of expensive liquid handling devices
  • Non-autoclavable
  • Available with sterile and non-sterile options
Specifications
Specifications
Polyethylene
For washing plates, feeding, and supernatant aspiration from microwell plates
Individually Wrapped
600 μL
Transfer Pipet
Sterile
100/Cs.
Natural
180 mm
Single-use
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ Transpette™ Eight Channel Transfer Pipettor > Sterile; 100/Pk.