Learn More
Thermo Scientific™ 96 Well Black/Clear Bottom Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10
Color:Black, Clear Bottom; Sterility; Non-sterile; Lid, Without Lid; Type: Optical Polymer Base
Brand: Thermo Scientific™ 265301
328.68 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Thermo Scientific™ Nunc MicroWell 96-Well Optical-Bottom Plates with Polymer Base have a solid polystyrene black or white upper structures with a polystyrene film at the bottom of the plate. The plates are optimized for fluorescence and luminescence in cell culture and microscopic applications.
White or black polystyrene upper structure with polymer or coverglass bottom plates combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications.
- Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
- Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
- Available in black for fluorescence assays or white for luminescence and fluorescence assays; both with clear bottoms
- Polymer base plate
- Working range, 50-200μL/well
- Customized bar coding is available
Ordering Information:
Part No. 152040, 152036, 152028 and 152037 is produced in a clean environment.

Specifications
Custom Barcoding Available | |
Without Lid | |
10 Pk., 30 Cases | |
Black | |
Polystyrene | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
10 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
8 x 12 | |
Nunc | |
Optical Polymer Base | |
400 μL | |
50 to 200 μL | |
Non-treated | |
30 |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1423031 | Certificate of Compliance | 21/02/2025 | 265301 |
1422467 | Certificate of Compliance | 20/02/2025 | 265301 |
1422466 | Certificate of Compliance | 18/02/2025 | 265301 |
1286138 | Certificate of Compliance | 11/12/2024 | 265301 |
1376052 | Certificate of Compliance | 11/12/2024 | 265301 |
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
42Hours
23Minutes
46Seconds
1 -
Multiple Manufacturers
More Discovery, Less Cost
Save on thousands of eligible products.Promo Code: 24111Expires: 2025-03-21
Days
2Hours
23Minutes
46Seconds
1New Offer Save Now -
Multiple Manufacturers
Grow Your Discovery
Fall in love with big savingsPromo Code: 24107Expires: 2025-03-28
Days
9Hours
23Minutes
46Seconds
1
Your input is important to us. Please complete this form to provide feedback related to the content on this product.