Learn More
Cytiva Benchkote Sheet for ÄKTA Systems
Protects the top buffer trays from buffer spillages and salt deposits
Brand:  Cytiva 2300-10060
 View more versions of this product
Product Code. 16062032
  • £19.40 / Pack of 10
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cover the top buffer tray to protect from buffer spillages and salt deposits
  • Simplify daily and weekly preventive maintenance of the top buffer tray of ÄKTA systems
  • Manufactured of absorbent, impermeable material that is designed to protect lab surfaces against hazardous spills
  • Smooth, absorbent Whatman paper quickly absorbs spills
  • Laminated polyethylene layer prevents flow-through to the working surface
  • Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
  • Convenient and easy to use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Absorbent Liner
Paper, Laminated Polyethylene
ÄKTA Pure
10/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Benchkote Sheet for ÄKTA Systems > ÄKTA Pure, 10/Pk