missing translation for 'onlineSavingsMsg'
Learn More
Benzoxazole, 97+%, Thermo Scientific Chemicals Product Code.: 10407772

Benzoxazole, 97+%, Thermo Scientific Chemicals

Product Code. 10407772
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 10407772

Brand: Thermo Scientific Acros 105680050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 273-53-0
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
MDL Number MFCD00005765
InChI Key BCMCBBGGLRIHSE-UHFFFAOYSA-N
Synonym benzoxazole, 1-oxa-3-azaindene, 1-oxa-3-aza-1h-indene, benzo d oxazole, benzooxazole, usaf ek-5017, unii-j233y1i55i, benzoxazol, benzoxazoline, benzothiazoieShow MoreShow Less
PubChem CID 9228
ChEBI CHEBI:38814
IUPAC Name 1,3-benzoxazole
SMILES O1C=NC2=CC=CC=C12

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 27°C to 30°C
Color Brown-Orange to Yellow
Boiling Point 182°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 27, 42
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,sulfuric acid
Formula Weight 119.12
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Benzoxazole, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0408069 Certificate TypeCertificate of Analysis Date02/05/2024 Catalog Number105680250,105681000,105680050
Lot NumberA0450826 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number105680250,105681000,105680050
compliance-icons compliance-icons
Product Identifier
  • Benzoxazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzoxazole, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.