missing translation for 'onlineSavingsMsg'
Learn More
Benzoyl chloride, 99.5+%, pure, Thermo Scientific Chemicals Product Code.: 10697242

Benzoyl chloride, 99.5+%, pure, Thermo Scientific Chemicals

Product Code. 10697242
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10697242

Brand: Thermo Scientific Acros 105750010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Benzoyl Chloride, 99%, C7H5ClO, CAS Number-98-88-4, unii-vty8706w36, a-chlorobenzaldehyde, benzaldehyde, alpha-chloro, benzoic acid chloride, benzenecarbonyl chloride, benzoic acid, chloride, alpha-chlorobenzaldehyde, benzoylchloride, ccris 802, benzoylchlorid, 2.5L, 98.5% min. (GC), 09, 182

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-88-4
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
MDL Number MFCD00000653
InChI Key PASDCCFISLVPSO-UHFFFAOYSA-N
Synonym benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36Show MoreShow Less
PubChem CID 7412
ChEBI CHEBI:82275
IUPAC Name benzoyl chloride
SMILES C1=CC=C(C=C1)C(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -1.0°C
Density 1.2100g/mL
Boiling Point 198.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula C6H5COCl
Refractive Index 1.5520 to 1.5540
Quantity 1 L
UN Number 1736
Beilstein 09, 182
Fieser 01,50; 05,24; 13,219
Merck Index 15, 1115
Specific Gravity 1.21
Solubility Information Solubility in water: reacts, 2 mg/mL. Other solubilities: decomposes in alcohols and in water, soluble in ether, benzene; miscible with oils,, carbon disulfide, ccl4
Viscosity 0.0012 Pas (30°C)
Formula Weight 140.57
Percent Purity 99.5%
Grade Pure
Physical Form Liquid
Chemical Name or Material Benzoyl Chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467463 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number105750025,105750010,10575090
Lot NumberA0461662 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number105750025,105750010,10575090
Lot NumberA0444416 Certificate TypeCertificate of Analysis Date22/01/2024 Catalog Number105750025,105750010,10575090
Lot NumberA0432127 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number105750025,105750010,10575090
Lot NumberA0395528 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number105750025,105750010,10575090
compliance-icons compliance-icons
Product Identifier
  • Benzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzoyl chloride, 99.5+%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.