missing translation for 'onlineSavingsMsg'
Learn More
beta-D-Glucose pentaacetate, 98%, Thermo Scientific Chemicals Product Code.: 11474184

beta-D-Glucose pentaacetate, 98%, Thermo Scientific Chemicals

Product Code. 11474184
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11474184

Brand: Thermo Scientific Alfa Aesar A12526.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

β-D-Glucose Pentaacetate is used in biochemical reaction and also used as an active pharmaceutical intermediate. Further, it is used to stimulate insulin release in rat pancreatic islets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-D-Glucose Pentaacetate is used in biochemical reaction and also used as an active pharmaceutical intermediate. Further, it is used to stimulate insulin release in rat pancreatic islets.

Solubility
Soluble in chloroform and methanol. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-69-3
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD00006597
InChI Key LPTITAGPBXDDGR-IBEHDNSVSA-N
Synonym beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-βShow MoreShow Less
PubChem CID 2724702
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 130°C to 132°C
Quantity 250 g
Beilstein 98851
Solubility Information Soluble in chloroform and methanol. Insoluble in water.
Optical Rotation +5° (c=5 in Chloroform)
Formula Weight 390.34
Percent Purity 98%
Chemical Name or Material beta-D-Glucose pentaacetate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252445 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog NumberA12526.18,A12526.0B,A12526.30
Lot Number10246969 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA12526.18,A12526.0B,A12526.30
Lot Number10171763 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12526.18,A12526.0B,A12526.30
Lot Number10173834 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12526.18,A12526.0B,A12526.30
Lot NumberX20G011 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12526.18,A12526.0B,A12526.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
beta-D-Glucose pentaacetate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.