Learn More
Corning™ BioCoat™ Endothelial Cell Growth Environment Kit
Increases endothelial cell yield from a variety of sources
Brand: Corning™ 355053
Includes: Complete Environment Kit: 500mL BD Endothelial Cell Culture Medium (MCDB 131 medium containing hydrocortisone, heparin and 2% fetal bovine serum; 5μg lyophilized Mouse Epidermal Growth Factor (EGF), receptor grade; 100mg lyophilized Bovine Endothelial Cell Growth Supplement (ECGS); 50mg lyophilized Soybean Trypsin Inhibitor; Five BD BioCoat Type I Collagen 75cm2 Flasks
Description
Ideal for human umbilical vein (HUVEC), pulmonary artery (HPAEC), and aorta (HAEC), as well as fetal bovine heart (FBHEC) cells. Interactions between components of the extracellular matrix and growth factors play a crucial role in determining the adaptation and responses of mammalian cells to their micro environments both in vivo and in vitro.
- Cells can be subcultured according to routine procedures, seeded into fermentation systems, or used in a variety of bioassays
- Integrate specially formulated low-serum or serum-free medium, culture supplements and growth factors
- Each environment lot is tested in a bioassay and all components are tested for bacteria and fungi
These products are for research use only and are not intended for diagnostic purposes.
Construction of model systems to study angiogenesis, atherosclerosis, wound healing and in vitro toxicology
Order Info
Media and cellware are also available separately.
Specifications
Complete Endothelial Cell Growth Environment Kit | |
Construction of model systems to study angiogenesis, atherosclerosis, wound healing and in vitro toxicology | |
2° to 8°C |
Product Suggestions
Customers who viewed this item also viewed