missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Biodyne™ B Membrane
Biodyne B membrane has pore surfaces populated by a high density of quaternary ammonium groups, making it strongly cationic.
Brand: Cytiva (formerly Pall Lab)
353.40 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.
This item is not returnable.
View return policy
Description
- This positive charge is maintained over a broad pH range and promotes strong ionic binding of nucleic acids, making it ideal for transfer and detection of nucleic acids
Specifications
Sheets | |
Positively-charged Nylon 6.6 | |
20 x 20 cm |
Non-sterile | |
0.45 μm | |
5.7 to 6.7 μm |