missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Biohazard Disposal Bags

Biohazard Disposal Bags for personal or general lab use, multiple sizes available to suit all needs.
£34.40 - £367.72
Product Code | Brand | Description | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Capacity (Metric) | Price | Quantity & Availability | |||||
15519083
![]() |
Heathrow Scientific™
HS10320 |
Biohazard Disposal Bags | 2 L |
£135.74
Pack of 500 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
16320676
![]() |
Heathrow Scientific™
HS10321 |
Biohazard Disposal Bags | 9 L |
£114.40
Pack of 200 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
|||||
15529083
![]() |
Heathrow Scientific™
HS10322 |
Biohazard Disposal Bags | 19 L |
£132.00
Pack of 200 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
16330676
![]() |
Heathrow Scientific™
HS10323 |
Biohazard Disposal Bags | 40 L |
£367.72
Pack of 200 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
|||||
16300676
|
Heathrow Scientific™
HS10044H |
Biohazard Disposal Bag Holder for HS10320 ONLY | 2 L |
£34.40
Each |
In Stock
Log in to see stock. ![]() |
|||||
Description
- Biohazard collection bag
- Biohazard warning symbol and precautions printed in English, Spanish, and French
- Includes an indicator patch that darkens when autoclaved
- Autoclavable to 250°F (121°C)
- Disposable
- HS10044H bag holder is ONLY for use with HS10320 2 L bag
Specifications
Autoclavable | |
Red | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Heathrow Scientific™ Biohazard Disposal Bags