missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Biohazard Disposal Bags

Biohazard Disposal Bags for personal or general lab use, multiple sizes available to suit all needs.

Brand:  Heathrow Scientific™ HS10044H

25.79 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 16300676

  • £34.40 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Biohazard collection bag
  • Biohazard warning symbol and precautions printed in English, Spanish, and French
  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 250°F (121°C)
  • Disposable
  • HS10044H bag holder is ONLY for use with HS10320 2 L bag
Specifications

Specifications

Bag Holder
Autoclavable
2 L
Reusable
HS10320 Biohazard Disposal Bag ONLY
Non-sterile
Shrink Wrap
3.6 x 5 x 8.4 in.
9.1 cm
12.8 cm
21.3 cm
0.16 kg
Biohazard Disposal Bag Holder for HS10320 ONLY
0.5 gal.
Red
Biohazard Disposal Bags (8 x 12 in.)
HDPE-coated Steel Wire
24 Pk.
3.6 x 5 in.
3.6 in.
5 in.
8.4 in.
0.36 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heathrow Scientific™ Biohazard Disposal Bags > Biohazard Disposal Bag Holder, One Biohazard Disposal Bag, Capacity: 2 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.