Learn More
Integra Biosciences™ 8-Channel Adapter with Ejector
Use this adapter with VACUSIP portable aspirators. Integra Biosciences™ 8-Channel Aspiration Adapters allow VACUSIP aspirators to provide solutions for a wide array of liquid waste disposal tasks.
Brand: Integra Biosciences™ 159024
This item is not returnable.
View return policy
Description
- 8-channel adapter with ejector for disposable tips
- For VACUSIP portable aspirators
- Unique VACUSIP hand operator allows for fine regulation of the liquid flow and can be connected to various adapters
- Different adapters are available to match a multitude of laboratory containers
Specifications
8-channel Adapter with Ejector | |
VACUSIP Portable Aspirator |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Integra Biosciences™ 8-Channel Adapter with Ejector > For use with: Generic tip
Spot an opportunity for improvement?Share a Content Correction