Learn More
Agilent BioTek Accessories For MultiFlo FX Microplate Dispenser RAD peri cassette
Accurate and precise peristaltic pump dispensing.
Brand:  Agilent BioTek 1260015
641.25 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
 View more versions of this product
Product Code. 16572092
  • £675.00 / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with MultiFlo FX Microplate Dispenser RAD peri cassette
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 to 3000 μL
MultiFlo FX Microplate Dispenser
10 μL
Plastic
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Agilent BioTek Accessories For MultiFlo FX Microplate Dispenser RAD peri cassette > Plastic tip; 10μ to 3000μL; 10μL increments