Learn More
Cytiva BioTrace™ NT Nitrocellulose Transfer Membrane
BioTrace NT is a pure unsupported nitrocellulose membrane that has high binding capacity for proteins and nucleic acids.
Brand:  Cytiva 66489
 View more versions of this product
Product Code. 17134571
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • High tensile strength makes it less likely to tear or crack than other brands of nitrocellulose
  • High binding capacity for both proteins and nucleic acids
  • Ideal for colony/plaque lifts and protein transfers
  • Exclusive dot grid pattern does not enhance or inhibit colony growth certified for use in the Membrane Filter (MF) Technique
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sheets
BioTrace NT
20 x 20 cm
Non-sterile
0.2 μm
101.6 to 190.5 μm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva BioTrace™ NT Nitrocellulose Transfer Membrane > 20 x 20 cm Sheets