missing translation for 'onlineSavingsMsg'
Learn More

Pall BioTrace™ NT Nitrocellulose Transfer Membrane

BioTrace NT is a pure unsupported nitrocellulose membrane that has high binding capacity for proteins and nucleic acids.

£140.00 - £349.00

Specifications

Description Non-sterile
Material BioTrace NT
Pore Size 0.2 μm
Thickness 101.6 to 190.5 μm
View More Specs

Products 3
Product Code Brand Dimensions (LxW) Price Quantity & Availability  
Product Code Brand Dimensions (LxW) Price Quantity & Availability  
17124571
View Documents
Pall
66485
30 cm x 3 m This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17134571
View Documents
Pall
66489
20 x 20 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17164581
View Documents
Pall
66593
7 x 8.5 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • High tensile strength makes it less likely to tear or crack than other brands of nitrocellulose
  • High binding capacity for both proteins and nucleic acids
  • Ideal for colony/plaque lifts and protein transfers
  • Exclusive dot grid pattern does not enhance or inhibit colony growth certified for use in the Membrane Filter (MF) Technique
Specifications

Specifications

Non-sterile
BioTrace NT
0.2 μm
101.6 to 190.5 μm
Videos
SDS
Documents

Documents

Product Certifications