missing translation for 'onlineSavingsMsg'
Learn More
Biphenyl-3,3',5,5'-tetracarboxylic acid, Thermo Scientific Chemicals Product Code.: 16677042

Biphenyl-3,3',5,5'-tetracarboxylic acid, Thermo Scientific Chemicals

Product Code. 16677042
100 mg
Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy

Product Code. 16677042

Brand: Thermo Scientific Acros 465831000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4371-28-2
Molecular Formula C16H10O8
Molecular Weight (g/mol) 330.25
MDL Number MFCD16621475
InChI Key QURGMSIQFRADOZ-UHFFFAOYSA-N
IUPAC Name [1,1'-biphenyl]-3,3',5,5'-tetracarboxylic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(O)=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O

Specifications

Alpha Vector BIPHENYLTETRACARBOXYLICACID
Melting Point >400°C
Color Brown to White
Infrared Spectrum Conforms
Assay Percent Range 94.0 to 106.0 % (Titration with NaOH)
Packaging glass bottle
Quantity 100 mg
UN Number 3077
Formula Weight 330.25
Physical Form Crystalline Solid or Powder
Chemical Name or Material Biphenyl-3,3 ',5,5 '-tetracarboxylic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459984 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number465831000,465830010
Lot NumberA0458023 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number465831000,465830010
Lot NumberA0416305 Certificate TypeCertificate of Analysis Date05/09/2022 Catalog Number465831000,465830010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Biphenyl-3,3',5,5'-tetracarboxylic acid, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.