missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BIRB 796
High affinity and selective p38 kinase inhibitor
£124.00 - £420.00
Specifications
Chemical Name or Material | N-[3-(1,1-Dimethylethyl)-1-(4-methylphenyl)-1H-pyrazol-5-yl]-N′-[4-[2-(4-morpholinyl)ethoxy]-1-naphthalenyl]urea |
---|---|
CAS | 285983-48-4 |
Synonym | Doramapimod |
Target | p38 Inhibitors |
Molecular Formula | C31H37N5O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18757996
|
Tocris Bioscience™
5989/10 |
10 mg |
£124.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18737196
|
Tocris Bioscience™
5989/50 |
50 mg |
£420.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
High affinity and selective p38 kinase inhibitor (Kd = 50-100 pM). Exhibits no significant inhibition on a panel of related kinases. Inhibits LPS-induced TNFα production in human PBMCs and whole blood (IC50 values are 21 and 960 nM, respectively). Also inhibits JNK2α2 and c-Raf-1 (IC50 values are 98 nM and 1.4 μM, respectively). Cell permeable.Specifications
N-[3-(1,1-Dimethylethyl)-1-(4-methylphenyl)-1H-pyrazol-5-yl]-N′-[4-[2-(4-morpholinyl)ethoxy]-1-naphthalenyl]urea | |
Doramapimod | |
C31H37N5O3 | |
0.98 |
285983-48-4 | |
p38 Inhibitors | |
527.66 |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title