Learn More
Bismuth carbonate oxide, 98.5% min, Thermo Scientific Chemicals
Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazes
£56.00 - £202.00
Chemical Identifiers
CAS | 5892-10-4 |
---|---|
Molecular Formula | CBi2O5 |
Molecular Weight (g/mol) | 509.97 |
MDL Number | MFCD00010877 |
InChI Key | MGLUJXPJRXTKJM-UHFFFAOYSA-L |
Synonym | bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide |
PubChem CID | 16683095 |
ChEBI | CHEBI:31291 |
IUPAC Name | bis(oxobismuthanyl) carbonate |
SMILES | C(=O)(O[Bi]=O)O[Bi]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327528
|
Thermo Scientific Alfa Aesar
033216.22 |
100 g |
£56.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337528
|
Thermo Scientific Alfa Aesar
033216.36 |
500 g |
£202.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazesBismuth carbonate oxide is used in ceramic glazes and pearly coatings on plastics. It is also used as an antacid and anti-diarrheal in medicinal chemistry. Further, it is used as a filler in radiopaque catheters. It is also employed in fireworks and to make Dragon's eggs. In addition to this, it is made into nanotube arrays that exhibit antibacterial properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5892-10-4 | |
509.97 | |
MGLUJXPJRXTKJM-UHFFFAOYSA-L | |
16683095 | |
bis(oxobismuthanyl) carbonate |
CBi2O5 | |
MFCD00010877 | |
bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide | |
CHEBI:31291 | |
C(=O)(O[Bi]=O)O[Bi]=O |
Specifications
5892-10-4 | |
CBi2O5 | |
14,1282 | |
Practically insoluble in water,alcohol; Soluble in nitric acid,hydrochloric acid,concentrated acetic acid;Soluble in mineral acid and glacial acetic acid; Insoluble in water. | |
C(=O)(O[Bi]=O)O[Bi]=O | |
509.97 | |
CHEBI:31291 | |
≤98.5% | |
Bismuth carbonate oxide |
Powder | |
MFCD00010877 | |
bismuth subcarbonate, basic bismuth carbonate, dibismuth carbonate dioxide, bismuth oxycarbonate, bismuth iii carbonate, 2,4-dioxa-1,5-dibismapentane, 1,3,5-trioxo, bismuth carbonate oxide bi2 co3 o2, 1,3,5-trioxo-2,4-dioxa-1,5-dibismapentane, bismuthyl carbonate, bismuth carbonate oxide | |
MGLUJXPJRXTKJM-UHFFFAOYSA-L | |
bis(oxobismuthanyl) carbonate | |
16683095 | |
509.97 | |
6.86 g/mL |
Safety and Handling
missing translation for 'einecsNumber' : 227-567-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only