Learn More
Bismuth carbonate oxide, 98.5% min, Thermo Scientific Chemicals
Description
Opacifier in X-ray diagnosis, enamel fluxes, ceramic glazesBismuth carbonate oxide is used in ceramic glazes and pearly coatings on plastics. It is also used as an antacid and anti-diarrheal in medicinal chemistry. Further, it is used as a filler in radiopaque catheters. It is also employed in fireworks and to make Dragon's eggs. In addition to this, it is made into nanotube arrays that exhibit antibacterial properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder |
Quantity | 500 g |
Merck Index | 14,1282 |
Solubility Information | Practically insoluble in water,alcohol; Soluble in nitric acid,hydrochloric acid,concentrated acetic acid;Soluble in mineral acid and glacial acetic acid; Insoluble in water. |
Formula Weight | 509.97 |
Percent Purity | ≤98.5% |
Density | 6.86 g/mL |
Chemical Name or Material | Bismuth carbonate oxide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S13K016 | Certificate of Analysis | 28/06/2024 | 033216.36,033216.22 |
S04K034 | Certificate of Analysis | 13/06/2024 | 033216.36,033216.22 |
N13J027 | Certificate of Analysis | 24/10/2023 | 033216.36,033216.22 |
W01J005 | Certificate of Analysis | 21/09/2023 | 033216.36,033216.22 |
M03A013 | Certificate of Analysis | 09/02/2023 | 033216.36,033216.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.