Learn More
Bismuth citrate, 94%, Thermo Scientific Chemicals
Description
Bismuth citrate is used as a progressive hair dye in cosmetics. It is used as antimicrobial agents for syphills, colitis, wound infection, quartan malaria, dyspepsia and peptic ulcers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | (decomposition) |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 250 g |
Solubility Information | Insoluble in water,alcohol,ether; Soluble in ammonia solution and in solution of alkali citrates; Typical tap density 0;50g/cm3;Soluble in ammonia,alkali citrates; Insoluble in water,alcohol and ether. |
Formula Weight | 398.08 |
Percent Purity | 94% |
Odor | Odorless |
Sensitivity | Light sensitive |
Density | 3.5 g/mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q02G025 | Certificate of Analysis | 09/07/2024 | 040398.30,040398.A1 |
U05I011 | Certificate of Analysis | 10/01/2023 | 040398.30,040398.A1 |
Y22H013 | Certificate of Analysis | 12/04/2022 | 040398.30,040398.A1 |
Y09D040 | Certificate of Analysis | 08/12/2021 | 040398.30,040398.A1 |
B28X028 | Certificate of Analysis | 07/12/2021 | 040398.30,040398.A1 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.