missing translation for 'onlineSavingsMsg'
Learn More
Bismuth(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11393317

Bismuth(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11393317
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11393317

Brand: Thermo Scientific Alfa Aesar 017115.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Catalysts for organic reactions, preparation of Bi salts.Bismuth(III) chloride is used as a catalyst in organic synthesis and to prepare other bismuth salts. It is used as a starting material for the preparation of bismuthoxychloride upon hydrolysis. It is an oxidizing agent to prepare metallic bismuth by reducing agent. Further, it catalyzes the Michael reaction and the Mukaiyama-aldol reaction. In addition to this, it is a Lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-60-2
MDL Number MFCD00003461

Specifications

Boiling Point ca 430°C (sublimation)
Melting Point 230°C to 232°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.99% (Metals basis)
Molecular Formula BiCl3
Solubility Information Soluble in HCl,HNO3,acetone,ethyl acetate. Insoluble in alcohol. Decomposed by water to BiOCl.Soluble in hydrochloric acid,nitric acid,diethyl ether,ethyl acetate and acetone. Insoluble in alcohol.
Formula Weight 315.34
Odor Irritating
Sensitivity Hygroscopic
Density 4.75
Chemical Name or Material Bismuth(III) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP18L004 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number017115.22,017115.14
Lot NumberQ02K053 Certificate TypeCertificate of Analysis Date09/04/2024 Catalog Number017115.22,017115.14
Lot NumberR10J022 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number017115.22,017115.14
Lot NumberW25J053 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog Number017115.22,017115.14
Lot NumberN01I021 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number017115.22,017115.14
compliance-icons
Product Identifier
  • Bismuth(III) chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.