Learn More
Bismuth(III) chloride oxide, 98%, Thermo Scientific Chemicals
Description
Bismuth(III) chloride oxide is used in pharmaceutical industry, in the production of cosmetics, in chemical and metallurgical industries. It is used as pigment, in manufacture of artificial pearls and as a dry-cell cathodes, in photocatalysts for degradation of organic pollutant, in white-light LEDs, in the manufacture of plastics, in the synthesis of methanol, in semiconductor devices such as CCDs or photodiodes and also used in the production of permanent magnets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | (decomposition) |
Color | Brown |
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | BiOCl |
Merck Index | 14,1263 |
Solubility Information | Insoluble in water,alcohol; Soluble in acids. |
Formula Weight | 260.43 |
Percent Purity | 98% |
Odor | Odorless |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
J16S062 | Certificate of Analysis | 08/12/2021 | 040396.30,040396.A1 |
K12S047 | Certificate of Analysis | 08/12/2021 | 040396.30,040396.A1 |
Q09B017 | Certificate of Analysis | 08/12/2021 | 040396.30,040396.A1 |
D18S039 | Certificate of Analysis | 07/12/2021 | 040396.30,040396.A1 |
D08P13 | Certificate of Analysis | 07/12/2021 | 040396.30,040396.A1 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.