missing translation for 'onlineSavingsMsg'
Learn More
Bismuth(III) oxide, typically 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11343327

Bismuth(III) oxide, typically 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11343327
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11343327

Brand: Thermo Scientific Alfa Aesar 017132.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as an adhesive and sealant chemical, flame retardant, pigment, propellant and blowing agent. It is also used as a solvent. Other uses are in batteries, electrical and electronic products, explosive materials, ink, toner, and colorant products, paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-76-3
Molecular Formula Bi2O3
Molecular Weight (g/mol) 465.96
MDL Number MFCD00003462
InChI Key TYIXMATWDRGMPF-UHFFFAOYSA-N
IUPAC Name dibismuth(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Bi+3].[Bi+3]

Specifications

Boiling Point ca 1890°C
Melting Point 817°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Insoluble in water. Soluble in acids.
Formula Weight 465.96
Density 8.9
Chemical Name or Material Bismuth(III) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD30M11 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number017132.18,017132.30
Lot NumberG03R009 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number017132.18,017132.30
Lot NumberT15A050 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number017132.18,017132.30
Lot NumberC01M16 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number017132.18,017132.30
Lot Number5001A04S Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number017132.18,017132.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth(III) oxide, typically 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.