missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bismuth(III) oxyfluoride, 97%, pure, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Specifications
Melting Point | 500.0°C |
Quantity | 100 g |
Assay Percent Range | 96% min. (ex Bi) |
Linear Formula | FBi=O |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 243.98 |
Percent Purity | 97% |
Grade | Pure |
Chemical Name or Material | Bismuth(III) oxyfluoride |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Safety and Handling


Product Identifier
- Bismuth(III) oxyfluoride
- Warning
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bismuth(III) oxyfluoride, 97%, pure, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction